Order Entry
Ireland
ContactUsLinkComponent
Pack of 1000units of empty 2mL reinforced Precellys tubes+caps. Precellys lysing kit cat. No. 03961-1-007 and 03961-1-008 are made w ith this reference. The 2mL reinforced tubes and caps are assembled, in PPCO with o-ring, autoclavable, validated for 1 * 1 KIT
Catalog # BERT0909-03
Supplier:  Bertin Instruments
Pack of 1000units of empty 2mL reinforced Precellys tubes+caps. Precellys lysing kit cat. No. 03961-1-007 and 03961-1-008 are made w ith this reference. The 2mL reinforced tubes and caps are assembled, in PPCO with o-ring, autoclavable, validated for 1 * 1 KIT
Catalog # BERT0909-03
Supplier:  Bertin Instruments
Frequently Bought Together