Order Entry
Ireland
ContactUsLinkComponent
Refrigerator, Premium Biofresh, total capacity 190L compartment-169L, energy Class-A++, 4-Star, height 1220 x width 1236 x depth 570 mm, Super Cool 1 * 1 items
Catalog # LIEBIKB2224
Supplier:  Liebherr
Refrigerator, Premium Biofresh, total capacity 190L compartment-169L, energy Class-A++, 4-Star, height 1220 x width 1236 x depth 570 mm, Super Cool 1 * 1 items
Catalog # LIEBIKB2224
Supplier:  Liebherr
Frequently Bought Together