Order Entry
Ireland
ContactUsLinkComponent
Magnetic stirrer, ultra flat, compact and strong with fully automatic stirring bar monitoring mixWATCH, up to 40 litres stirring vol ume, also suitable for highly viscous media, very strong magnetic coupling, therefore predestined for magnetic power 1 * 1 items
Catalog # TMAG40700_CH
Supplier:  2MAG
Magnetic stirrer, ultra flat, compact and strong with fully automatic stirring bar monitoring mixWATCH, up to 40 litres stirring vol ume, also suitable for highly viscous media, very strong magnetic coupling, therefore predestined for magnetic power 1 * 1 items
Catalog # TMAG40700_CH
Supplier:  2MAG
Frequently Bought Together