Order Entry
Ireland
ContactUsLinkComponent
Heating bath circulator steel, sc100-s3, amb+13 to 100 c, 3-5 l bath volume, 2.0 kw heater, pump 17 l/min & 300 mbar 3 years warra nty 230v/50hz 1 * 1 items
Catalog # HAKA152-1031
Heating bath circulator steel, sc100-s3, amb+13 to 100 c, 3-5 l bath volume, 2.0 kw heater, pump 17 l/min & 300 mbar 3 years warra nty 230v/50hz 1 * 1 items
Catalog # HAKA152-1031
Frequently Bought Together