Order Entry
Ireland
ContactUsLinkComponent
Elmasonic X-tra basic 550 - ultrasonic cleaner Int.Dem. of tank (mm) 500x330x350, Volume: 58 liters, Serv. Cap. 45 1 * 1 items
Catalog # ELMA1026060
Supplier:  ELMA SCHMIDBAUER
Elmasonic X-tra basic 550 - ultrasonic cleaner Int.Dem. of tank (mm) 500x330x350, Volume: 58 liters, Serv. Cap. 45 1 * 1 items
Catalog # ELMA1026060
Supplier:  ELMA SCHMIDBAUER
Frequently Bought Together