Order Entry
Ireland
Orders LinkContactUsLinkComponent
VWR® Dip Slides

VWR® Dip Slides

TSA with malt extract agar, on both sides, for yeasts and moulds, 10
Catalog # 535320D
Supplier:   VWR Chemicals
VWR® Dip Slides
Catalog # 535320D
Supplier:  VWR Chemicals
CAS Number:  
VWR® Dip Slides
VWR® Dip Slides

TSA with malt extract agar, on both sides, for yeasts and moulds, 10

CAS Number:  
Price
19,80 €
Log in for availability
1 * 10 items
Product Details & Documents

Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.

  • Supplied sterile
  • Sampling takes only a few seconds
  • Results are easily read after just 24 to 48 hours
Specifications

Specifications

  • Pk:
    10
  • Description:
    TSA with malt extract agar, on both sides, for yeasts and moulds
  • Cat. No.:
    535320D
  • Type:
    Push cap
  • Paddle size:
    80×28 mm|80×28
  • Dimensions:
    30 mm tube, 110 mm high
  • Tray W×L:
    50×22 mm
  • Contact area:
    10 cm²|10

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.