0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
BOTTLE 2L ROUND 1 * 50 items
COVE5068
:
COVERA
BOTTLE 2L ROUND 1 * 50 items
COVE5068
:
COVERA
Frequently Bought Together
Orthophosphoric acid 85% ≥85%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Connection system GL 45
: SciLabware
Bottles, narrow neck, square, HDPE, with screw caps, LaboPlast®
: BURKLE
Genuine Tecan® 50 µL disposable automation tips designed specifically for use on the Freedom EVO® and Fluent® MultiChannel Arm™ (MC A) 96 option. ANSI format, single stack, filtered. Tecan STERILE purity level - sterile, tested and certified to b an 1 * 3.840 items
: Tecan
Accessories for container PACKO
: CURTEC
Microtube racks, floating, Nalgene®
: Thermo Fisher Scientific