Indophenol, Sigma-Aldrich®
5 g
Catalog # I5763-5G
Supplier: SIGMA ALDRICH MICROSCOPY
Indophenol, Sigma-Aldrich®
Catalog # I5763-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number: 500-85-6
Jump to:
Product Details & Documents
Indophenol is used in hair dyes, redox materials, lubricants, liquid crystal displays, biosensor and fuel cells. It is toxic to fishes and is implicated in environmental pollution. Indophenol method is common for the determination of ammonia. The reaction gives a blue product, which is measured spectrophotometrically.
Synonyms: Phenolindophenol
- CAS Number:500-85-6
- Formula:C₁₂H₉NO₂
- MDL Number:MFCD00001621
- Molecular Weight:199.21 g/mol
- Melting Pt:>300 °C (lit.)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Room Temperature
- EINECS:207-913-5
Specifications
Specification Test Results
- Appearance (Colour):Dark Green to Black
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Solubility (Colour):Blue to Dark Blue to Dark Green
- Solubility (Turbidity), 10 mg/ml, 1 M NaOH:Clear
Specifications
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



