Order Entry
Ireland
Orders LinkContactUsLinkComponent
Wire baskets for transport

Wire baskets for transport

Mesh dimensions: 35×35×3 mm, 300×400×150 mm
Catalog # BOCH17312
Supplier:   BOCHEM
Wire baskets for transport
Catalog # BOCH17312
Supplier:  BOCHEM
Wire baskets for transport
Wire baskets for transport

Mesh dimensions: 35×35×3 mm, 300×400×150 mm

Price
483,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Stainless steel 18/10.

  • Three different mesh dimensions
  • Stackable
  • Excellent thermal and chemical resistance

These rectangular baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.

Specifications
  • Description:
    Mesh dimensions: 35×35×3 mm
  • Seq:
    03
  • W×D×H:
    300×400×150 mm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • 275×375 mm, mesh dimensions: 35×35×3 mm
    275×375 mm, mesh dimensions: 35×35×3 mm
    BOCH17326
    Log in for availability
    1 * 1 items - 403,00 €

Product Information

  • Catalog #
    Availability
    Price
  • 575×375 mm, mesh dimensions: 35×35×3 mm
    575×375 mm, mesh dimensions: 35×35×3 mm
    BOCH17325
    Log in for availability
    1 * 1 items - 437,00 €

Product Information

  • Catalog #
    Availability
    Price
  • 275×375 mm, mesh dimensions: 20×20×3 mm
    275×375 mm, mesh dimensions: 20×20×3 mm
    BOCH17321
    Log in for availability
    1 * 1 items - 403,00 €

Product Information

  • Catalog #
    Availability
    Price
  • 575×375 mm, mesh dimensions: 20×20×3 mm
    575×375 mm, mesh dimensions: 20×20×3 mm
    BOCH17320
    Log in for availability
    1 * 1 items - 437,00 €

Product Information

  • Catalog #
    Availability
    Price
  • 275×375 mm, mesh dimensions: 10×10×3 mm
    275×375 mm, mesh dimensions: 10×10×3 mm
    BOCH17316
    Log in for availability
    1 * 1 items - 414,00 €

Product Information

  • Catalog #
    Availability
    Price
  • 575×375 mm, mesh dimensions: 10×10×3 mm
    575×375 mm, mesh dimensions: 10×10×3 mm
    BOCH17315
    Log in for availability
    1 * 1 items - 449,00 €