Order Entry
Ireland
Orders LinkContactUsLinkComponent
Palmitoleic acid

Palmitoleic acid

Glass bottle for solids, 5 g
Catalog # CAYM10009871-5
Supplier:   Cayman Chemical
Palmitoleic acid
Catalog # CAYM10009871-5
Supplier:  Cayman Chemical
CAS Number:  373-49-9
Palmitoleic acid

Glass bottle for solids, 5 g

CAS Number:  373-49-9
Price
1 100,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (Z)-Hexadec-9-enoic acid
  • CAS Number:373-49-9
  • Formula:C₁₆H₃₀O₂
  • MDL Number:MFCD00004437
  • Molecular Weight:254.41 g/mol
  • EINECS:206-765-9

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications