gas cylinder cabinet, single walled, medium cabinet with wing door(s), model line g-line, model class g-pg, model , version perforat ed dimensions: exterior: wxdxh (mm) 1680 x 690 x 1500 interior: wxdxh (mm) 1660 x 660 x 1470 1 * 1 items
Catalog # MSGA2846
Supplier: asecos
Frequently Bought Together