Order Entry
Ireland
ContactUsLinkComponent
perforated insert standard, sheet steel, plastic laminated plain, ral 7035 light grey load capacity (evenly distributed load) (kg): 75 for underbench cabinets with width 590/1100 t=500 mm for model(s): ub90 1 * 1 items
  MSGA18814
 :  asecos
perforated insert standard, sheet steel, plastic laminated plain, ral 7035 light grey load capacity (evenly distributed load) (kg): 75 for underbench cabinets with width 590/1100 t=500 mm for model(s): ub90 1 * 1 items
  MSGA18814
 :  asecos
Frequently Bought Together