Order Entry
Ireland
ContactUsLinkComponent
KIT CERAMIC 2,8MM REINFORCED 2 ML TUBES, ready-to-use kits with the Precellys24, includes 50 microtubes, prefilled with 2,8mm cera mic (zirconium oxide) beads in 2mL reinforced tubes.Custom design reinforced tubes for bead beating application : o-ri 1 * 1 KIT
Catalog # BERT03961-1-007
Supplier:  Bertin Instruments
KIT CERAMIC 2,8MM REINFORCED 2 ML TUBES, ready-to-use kits with the Precellys24, includes 50 microtubes, prefilled with 2,8mm cera mic (zirconium oxide) beads in 2mL reinforced tubes.Custom design reinforced tubes for bead beating application : o-ri 1 * 1 KIT
Catalog # BERT03961-1-007
Supplier:  Bertin Instruments
Frequently Bought Together