Order Entry
Ireland
Orders LinkContactUsLinkComponent
Microscope slides

Microscope slides

Microscope slides with frosted one end on one side, 1,0 - 1,2 mm, 76×26 mm
Catalog # SMITMC2952FB/76X26
Supplier:   SMITH SCIENTIFIC
Microscope slides
Catalog # SMITMC2952FB/76X26
Supplier:  SMITH SCIENTIFIC
Microscope slides

Microscope slides with frosted one end on one side, 1,0 - 1,2 mm, 76×26 mm

Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents

These slides are made of glass with ground edges.

Specifications
  • Description:
    Microscope slides with frosted one end on one side
  • L×W:
    76×26 mm
  • Seq:
    003
  • Thickness:
    1,0 - 1,2 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.