Order Entry
Ireland
Orders LinkContactUsLinkComponent
Büchner funnels
Büchner funnels
192 mm, 185 mm
Catalog # SMIT237-8
Supplier:   SMITH SCIENTIFIC
Büchner funnels
Catalog # SMIT237-8
Supplier:  SMITH SCIENTIFIC
Büchner funnels
Büchner funnels
192 mm, 185 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
In white glazed porcelain.
For paper or membrane filtration discs.
Specifications
  • Plate Ø:
    192 mm
  • For filter Ø:
    185 mm