Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
ISO 23696-1: Water Quality — Determination of Nitrate in Water Using Small-Scale Sealed Tubes — Part 1
• Method: This part specifies the nitrate determination using the Dimethyl phenol (DMP) reaction.
• Citation Forms: The primary citation form is NO₃-N, with NO₃⁻ as an alternative.
• Spectroquant® Cell Tests 1.00614, 1.14563, and 1.14764 align with this norm, employing the DMP method.
Documents
Certificates
Search by Lot NumberMerck Individual products and tailor-made chemicals for your technical applications Merck Pharma Segment Brochure Merck Segment Brochure
Product Literature
Test Kits, Nitrate, Spectroquant®, Supelco®
Additional Product Details
Specifications
Specifications
- Chemical method:2,6-Dimethylphenol
- Pk:250 Tests
- Description:Nitrate test
- For unit:Prove, Nova 60, Picco
- Range:0,10 - 25,0 mg/L NO₃ - N, 0,4 - 110,7 mg/L NO₃
- Cat. No.:1.09713.0002
- M-Clarity quality level:MQ100
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



