Order Entry
Ireland
ContactUsLinkComponent
COLUMN AMIDE-80, 5 UM, 80A - SILICA - 4.6MM ID X 5CM L 1 * 1 items
Catalog # TOYO0019532
Supplier:  TOSOH Bioscience
COLUMN AMIDE-80, 5 UM, 80A - SILICA - 4.6MM ID X 5CM L 1 * 1 items
Catalog # TOYO0019532
Supplier:  TOSOH Bioscience
Frequently Bought Together