Order Entry
Ireland
ContactUsLinkComponent
Insert, volume 5mm, colour clear, material polyspring glass, usable volume 175µl, residual volume <1µl, self aligning and provide a cushion against needle contact
Catalog # CRMAC4012-530
Supplier:  CHROMACOL
Insert, volume 5mm, colour clear, material polyspring glass, usable volume 175µl, residual volume <1µl, self aligning and provide a cushion against needle contact
Catalog # CRMAC4012-530
Supplier:  CHROMACOL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.