Order Entry
Ireland
ContactUsLinkComponent
Vial, capacity 2ml, target 10-425 wide opening screw thread, diameter 12mm, height 32mm, flat bottom, material silanized glass, colo ur amber, usable volume 1.5ml, patched, compatible with many non-robotic autosamplers
Catalog # CRMAC4010-S2W
Supplier:  CHROMACOL
Vial, capacity 2ml, target 10-425 wide opening screw thread, diameter 12mm, height 32mm, flat bottom, material silanized glass, colo ur amber, usable volume 1.5ml, patched, compatible with many non-robotic autosamplers
Catalog # CRMAC4010-S2W
Supplier:  CHROMACOL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.