Order Entry
Ireland
Orders LinkContactUsLinkComponent
Agarose, SeaKem® HEEO
Agarose, SeaKem® HEEO
Agarose, SeaKem® HEEO
Catalog # 733-1558
Supplier:   LONZA
Agarose, SeaKem® HEEO
Catalog # 733-1558
Supplier:  LONZA
Agarose, SeaKem® HEEO
Agarose, SeaKem® HEEO
Agarose, SeaKem® HEEO
Price
220,00 €
Log in for availability
1 * 25 g
Product Details & Documents
SeaKem® HEEO Agarose is a very high EEO agarose that is useful in applications requiring significant cathodal migration, such as IEP of IgG and IgM. It can also be used to blend with a lower EEO agarose to achieve a specific EEO value.
Documents
Specifications
  • Description:
    Agarose, SeaKem® HEEO
  • Seq:
    01