Containers, wide mouth, square with thread
2000 ml, 122,0×122,0×187,0 mm, 69,0 mm, 80 mm
216-3906
: Kautex
PETG. Excellent transparency. Containers are supplied without closure. Black closure with PE-foam wad must be ordered separately.
- Wide neck opening
- Easy to fill, easy to clean
- Leakproof
- Suitable for food
- Crystal-clear for an excellent product presentation
- Halal certified
This container is also suitable as a sales or gift packaging.
- Thread:80 mm
- Capacity:2000 ml
- W×D×H:122,0×122,0×187,0 mm
- Neck I-Ø:69,0 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



