Order Entry
Ireland
ContactUsLinkComponent
Kit protein dotMETRIC with spot application device (> 300 assays). Protein assay for rapid estimation of protein concentration with only a single micro liter of protein sample. Test strip is spotted and results are visible from 2-8 minutes with no pr 1 * 1 KIT
Catalog # 786-21
Supplier:  G-Biosciences
undefined
Kit protein dotMETRIC with spot application device (> 300 assays). Protein assay for rapid estimation of protein concentration with only a single micro liter of protein sample. Test strip is spotted and results are visible from 2-8 minutes with no pr 1 * 1 KIT
Catalog # 786-21
Supplier:  G-Biosciences