Kit protein dotMETRIC (> 300 assays). Protein assay for rapid estimation of protein concentration with only a single micro liter of protein sample. Test strip is spotted and results are visible from 2-8 minutes with no protein to protein variation. N 1 * 1 KIT
Catalog # 786-20
Supplier: G-Biosciences
Frequently Bought Together