REFRIGERATOR TOTAL CAPACITY-301L WITH FREEZER VOLUME-56L DIMENSIONS-HEIGHT-1612MM LENGTH-600MM WIDTH-632MM 1 * 1 items
LIEBCUPEX3153
: Liebherr
REFRIGERATOR TOTAL CAPACITY-301L WITH FREEZER VOLUME-56L DIMENSIONS-HEIGHT-1612MM LENGTH-600MM WIDTH-632MM 1 * 1 items
LIEBCUPEX3153
: Liebherr
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection