Order Entry
Ireland
ContactUsLinkComponent
Semi-automatic, electro mechanically controlled benchtop laboratory autoclave with UK-plug. Chamberdimension (xL) 500 x 750 mm Volu me: 160 liters 1 * 1 items
Catalog # 481-2163
Supplier:  TUTTNAUER
Semi-automatic, electro mechanically controlled benchtop laboratory autoclave with UK-plug. Chamberdimension (xL) 500 x 750 mm Volu me: 160 liters 1 * 1 items
Catalog # 481-2163
Supplier:  TUTTNAUER
Frequently Bought Together