Order Entry
Ireland
Orders LinkContactUsLinkComponent
Calcein AM

Calcein AM

10 x 50 µg, 500 µG
Catalog # 734-1434
Supplier:   Corning
Calcein AM
Catalog # 734-1434
Supplier:  Corning
CAS Number:  148504-34-1
Calcein AM
Calcein AM

10 x 50 µg, 500 µG

CAS Number:  148504-34-1
Price
377,00 €
Log in for availability
1 * 500 µG
Product Details & Documents

The fluorescent dye calcein acetoxymethylester (calcein AM) can be used to label cells when performing analyses such as tumour cell invasion, endothelial cell migration, endothelial cell tubulogenesis, and other cell-based assays.

  • Non fluorescent cell viability indicator
  • Cell permeant compound that is hydrolysed by intracellular esterases in the fluorescent anion calcein

Calcein AM can be used to fluorescently pre- and post-label viable cells and perform kinetic and endpoint experiments, respectively.

The fluorescent dyes, calcein acetoxymethylester (calcein AM), can be used with Falcon* Corning FluoroBlok* cell culture inserts and insert systems to label cells when performing analyses such as tumor cell invasion, endothelial cell migration, and other cell-based assays

Caution: NOTE: Cells are known to tolerate different fluorescent dyes for varying amounts of time in culture. Cells labelled with calcein AM should be exposed to the dye for less than eight hours.

Synonyms:  Calcein-AM, Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester, Calcein acetoxymethyl ester, Tetrakis(acetoxymethyl) 2,2',2'',2'''-(((3',6'-diacetoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetate
  • CAS Number:148504-34-1
  • Formula:C₄₆H₄₆N₂O₂₃
  • MDL Number:MFCD05861516
  • Molecular Weight:994.86 g/mol

Documents

Specifications

Specifications

  • Pk:
    500 µG
  • Pk Info:
    10 x 50 µg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.