Indophenol, Sigma-Aldrich®
1 g
I5763-1G
Indophenol is used in hair dyes, redox materials, lubricants, liquid crystal displays, biosensor and fuel cells. It is toxic to fishes and is implicated in environmental pollution. Indophenol method is common for the determination of ammonia. The reaction gives a blue product, which is measured spectrophotometrically.
: Phenolindophenol
- :500-85-6
- :C₁₂H₉NO₂
- :MFCD00001621
- :199.21 g/mol
- :>300 °C (lit.)
- :M-Clarity™ quality level = MQ100
- :Room Temperature
- :207-913-5
- Appearance (Colour):Dark Green to Black
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Solubility (Colour):Blue to Dark Blue to Dark Green
- Solubility (Turbidity), 10 mg/ml, 1 M NaOH:Clear
- Pk:1 g



