Quantitative filter papers
4, 150 mm
Catalog # F4/KA2/150
Supplier: SMITH SCIENTIFIC
Jump to:
Product Details & Documents
Professional filter paper grade 1 for routine laboratory applications and professional filter paper grade 4 for rapid filtration with excellent retention of larger particles.
Specifications
- Grade:4
- Seq:02
- Size:150 mm