Order Entry
Ireland
Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
19/26 NS, 19/26 NS
Catalog # 8SMAP/22
Supplier:   SMITH SCIENTIFIC
Multiple adapters
Catalog # 8SMAP/22
Supplier:  SMITH SCIENTIFIC
Multiple adapters
19/26 NS, 19/26 NS
Price
170,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Multiple neck, two parallel necks.
Specifications
  • Cone size:
    19/26 NS
  • Socket size:
    19/26 NS
  • Seq:
    03