Order Entry
Ireland
Orders LinkContactUsLinkComponent
Sealed cell assemblies
Permanently sealed omni-cell window units, Csl, 0,10 mm thickness
  1936.
 :   Specac
Sealed cell assemblies
  1936.
 :  Specac
Sealed cell assemblies
Permanently sealed omni-cell window units, Csl, 0,10 mm thickness
1 * 1 Each
Rectangular with lead spacer sealed cell assemblies for omni cell units.
  • Description:
    Permanently sealed omni-cell window units, Csl, 0,10 mm thickness