Sealed cell assemblies
Permanently sealed omni-cell window units, Silica (IR), 0,05 mm thickness
Catalog # 1928.
Supplier: Specac
Jump to:
Product Details & Documents
Rectangular with lead spacer sealed cell assemblies for omni cell units.
Specifications
- Description:Permanently sealed omni-cell window units, Silica (IR), 0,05 mm thickness