Order Entry
Ireland
Orders LinkContactUsLinkComponent
Sealed cell assemblies
Sealed cell assemblies
Permanently sealed omni-cell window units, BaF2, 0,05 mm thickness
Catalog # 1923.
Supplier:   Specac
Sealed cell assemblies
Catalog # 1923.
Supplier:  Specac
Sealed cell assemblies
Permanently sealed omni-cell window units, BaF2, 0,05 mm thickness
Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents
Rectangular with lead spacer sealed cell assemblies for omni cell units.
Specifications
  • Description:
    Permanently sealed omni-cell window units, BaF2, 0,05 mm thickness