Sodium octanoate 96%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 96% CAS No.: 1984-06-1 Molecular Formula: CH3(CH2)6CO2Na Formula Weight: 166.20 Melting Point: 300° Beilstein Reference: 3597723
Synonyms: Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate
- CAS Number:1984-06-1
- Formula:C₈H₁₅NaO₂
- MDL Number:MFCD00058511
- Molecular Weight:166.2 g/mol
- Melting Pt:>300 °C
- Density:>0,9949 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:217-850-5
Documents
Certificates
Search by Lot NumberALFAA13451.36_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



