Order Entry
Ireland
Orders LinkContactUsLinkComponent
Hydroxyurea ≥98%

Hydroxyurea ≥98%

Glass bottle for solids, 25 g
Catalog # A10831.14
Health
Hydroxyurea ≥98%
Catalog # A10831.14
CAS Number:  127-07-1
Hydroxyurea ≥98%
Hydroxyurea ≥98%

Glass bottle for solids, 25 g

CAS Number:  127-07-1
Price
378,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 127-07-1 Molecular Formula: CH4N2O2 Formula Weight: 76.06 Melting Point: ca 140° dec. Beilstein Reference: 1741548 Merck Reference: 14,4848 MDL No.: MFCD00007943

An inhibitor of DNA synthesis and an anticancer agent

Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:14,4848
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:∼ 140 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:204-821-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.