Viscosity Reference sample N10 1 * 500 mL
Catalog # PSLE2700-V06
Supplier: POULTEN SELFE LEE
Frequently Bought Together
Iodomethane stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Hydrochloric acid ≥36,5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Diethyl ether stabilised, BAKER, J.T.Baker®
Supplier: Avantor
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals