Order Entry
Ireland
ContactUsLinkComponent
Flask volumetric, capacity 100ml, class A, amber graduated, polystoppered, tolerance 0.1ml, height 170mm, NS-ground 12/21, graduatio n color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # 264018008.
Supplier:  HIRSCHMANN
Flask volumetric, capacity 100ml, class A, amber graduated, polystoppered, tolerance 0.1ml, height 170mm, NS-ground 12/21, graduatio n color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # 264018008.
Supplier:  HIRSCHMANN
Frequently Bought Together