Order Entry
Ireland
ContactUsLinkComponent
Flask volumetric, capacity 50ml, class A, amber graduated, glass stoppered, tolerance 0.06ml, height 140mm, NS-ground 12/21, graduat ion color white, conformity certified. 1 * 1 items
Catalog # HIRS2640275_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 50ml, class A, amber graduated, glass stoppered, tolerance 0.06ml, height 140mm, NS-ground 12/21, graduat ion color white, conformity certified. 1 * 1 items
Catalog # HIRS2640275_S
Supplier:  HIRSCHMANN
Frequently Bought Together