Ely/a,electrolyte for d.o. probes series trioxmatic® 600/601/690/700 (iq)/701 (iq)/702 (iq)included: 1 bottle with 50 ml 1 * 50 mL
Catalog # WTWO205212
Supplier: WTW ONLINE
Ely/a,electrolyte for d.o. probes series trioxmatic® 600/601/690/700 (iq)/701 (iq)/702 (iq)included: 1 bottle with 50 ml 1 * 50 mL
Catalog # WTWO205212
Supplier: WTW ONLINE
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pierce™ BCA™ Protein assay kits and reagents
Supplier: Thermo Fisher Scientific
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Sample container, with screw cap
Supplier: DELTALAB