Fractionating columns, Vigreux
300 mm, 29/32 NS, 29/32 NS
Catalog # 201-0178
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With or without glass jacket.
Specifications
- Cone size:29/32 NS
- Socket size:29/32 NS
- Total length:300 mm