Büchner funnels, Hartley pattern
125 mm
Catalog # 511-0833
Supplier: DIXON SCIENTIFIC
Jump to:
Product Details & Documents
Top and base: Borosilicate glass with support plate.
- Available with choice of perforated disc material
- Complete with three PE coated clips
- Available for 4 filter sizes
Three piece construction.
Specifications
- For filter Ø:125 mm