Order Entry
Ireland
Orders LinkContactUsLinkComponent
Büchner funnels, Hartley pattern
Büchner funnels, Hartley pattern
70 mm
Catalog # 511-0827
Supplier:   DIXON SCIENTIFIC
Büchner funnels, Hartley pattern
Catalog # 511-0827
Supplier:  DIXON SCIENTIFIC
Büchner funnels, Hartley pattern
Büchner funnels, Hartley pattern
70 mm
Price
321,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Top and base: Borosilicate glass with support plate.
  • Available with choice of perforated disc material
  • Complete with three PE coated clips
  • Available for 4 filter sizes
Three piece construction.
Specifications
  • For filter Ø:
    70 mm