Order Entry
Ireland
Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
100 ml, 19/26 NS
Catalog # 221-0067
Supplier:   JONES FR
Conical separating funnels
Catalog # 221-0067
Supplier:  JONES FR
Conical separating funnels
Conical separating funnels
100 ml, 19/26 NS
Price
99,80 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass, with NS-stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml