Order Entry
Ireland
ContactUsLinkComponent
Wiper,sterile,70% 2-propanol,30% deionized water,Prosa
Catalog # 115-0902
Supplier:  CONTEC
Wiper,sterile,70% 2-propanol,30% deionized water,Prosa
Catalog # 115-0902
Supplier:  CONTEC