Order Entry
Ireland
ContactUsLinkComponent
RFM840 DIGITAL REFRACTOMETER 1.32-1.58/0.00001RI, 0-100/0.01 BRIX, PELTIER TEMPERATURE CONTROL 1 * 1 items
Catalog # 635-0369
Supplier:  BELLINGHAM STANLEY
RFM840 DIGITAL REFRACTOMETER 1.32-1.58/0.00001RI, 0-100/0.01 BRIX, PELTIER TEMPERATURE CONTROL 1 * 1 items
Catalog # 635-0369
Supplier:  BELLINGHAM STANLEY
Frequently Bought Together