Order Entry
Ireland
ContactUsLinkComponent
Bottle, reagent, capacity 100ml, diameter 54mm, height 100mm, narrow neck, with standard ground joint, NS 14/23, clear, soda lime gl ass 1 * 10 items
Catalog # SCOT231642407
Supplier:  DWK Life Sciences
Bottle, reagent, capacity 100ml, diameter 54mm, height 100mm, narrow neck, with standard ground joint, NS 14/23, clear, soda lime gl ass 1 * 10 items
Catalog # SCOT231642407
Supplier:  DWK Life Sciences
Frequently Bought Together