Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Saybolt colour -10 Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0044
Supplier:
Lovibond Tintometer
Expand image
Saybolt colour -10 Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0044
Supplier:
Lovibond Tintometer
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Magnesium standard solution, 1000 mg/L Mg in 0,5 M nitric acid (from Mg(NO₃)₂) Certipur® standard for AAS, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Conductivity standards, potassium chloride, referenced at 25 °C
Supplier: Hach