Bunsen burners
Bunsen burner, with air regulation, Natural gas
Catalog # RSGA253.030
Supplier: RSG
Jump to:
Product Details & Documents
Bunsen burner made of stainless steel.
- With air regulation sleeve
Specifications
- Description:Bunsen burner, with air regulation
- For:Natural gas