Gardner colour 8 Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0062
Supplier: Lovibond Tintometer
Gardner colour 8 Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0062
Supplier: Lovibond Tintometer
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Turbidity standards, Stablcal®
Supplier: Hach