Order Entry
Ireland
Orders LinkContactUsLinkComponent
Flanged fingertight nuts
Flanged fingertight nuts
PP washer for 1/16" flanged fittings
Catalog # 554-0750
Supplier:   VICI JOUR RESEARCH
Flanged fingertight nuts
Catalog # 554-0750
Supplier:  VICI JOUR RESEARCH
Flanged fingertight nuts
Flanged fingertight nuts
PP washer for 1/16" flanged fittings
Price
18,10 €
Log in for availability
1 * 10 items
Product Details & Documents
Fiberglass reinforced PP with PP washers. A number of chromatography systems still require or prefer flanged connections. When the tubing is flanged, the washer never comes in contact with the mobile phase.
  • Compatible with all 1/4"-28 flat-bottom ports
  • Available for 1/16" and 1/8" tubing
Pressure rating: Up to 30 bar (435 psi).
Pressure rating: Up to 30 bar (435 psi).
Specifications
  • Description:
    PP washer for 1/16" flanged fittings
  • Seq:
    2