Tweezer 120mm round tip straight, curved or tapered d=10mm n=1.5mm p=60mm r=45mm s=35mm u=8mm 1 * 1 items
Catalog # 232-1240
Supplier: OUTILS RUBIS
Tweezer 120mm round tip straight, curved or tapered d=10mm n=1.5mm p=60mm r=45mm s=35mm u=8mm 1 * 1 items
Catalog # 232-1240
Supplier: OUTILS RUBIS
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Calibration Weights, Class E2
Supplier: VWR Collection
pH/mV/°C meters, bench, inoLab® pH 7110 / 7310
Supplier: WTW
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Men's Coats, Cotton or Polyester/Cotton
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection